3-Chloro-4-methylanisole 97%
Dodavatel: Apollo Scientific
Synonyms:
2-Chloro-4-methoxy-1-methylbenzene, 3-Chloro-4-methylanisole, 2-Chloro-4-methoxytoluene
OR480614-100G
OR480614-25G
APOSOR480614-100GEA
6350
CZK
APOSOR480614-100G
APOSOR480614-25G
3-Chloro-4-methylanisole 97%
3-Chloro-4-methylanisole
Vzorec:
C₈H₉ClO MW: 156,61 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00087376 CAS číslo: 54788-38-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...