Tetrabromo-p-benzoquinone 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2,3,5,6-Tetrabromo-1,4-benzoquinone, 2,3,5,6-Tetrabromocyclohexa-2,5-diene-1,4-dione, p-Bromanil, Tetrabromoquinone, 4-Bromanil, Tetrabromo-1,4-benzoquinone
OR480592-25G
OR480592-100G
APOSOR480592-25GEA
4600
CZK
APOSOR480592-25G
APOSOR480592-100G
Tetrabromo-p-benzoquinone 95%
Tetrabromo-p-benzoquinone
Vzorec:
C₆Br₄O₂ MW: 423,68 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00013785 CAS číslo: 488-48-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...