Tisk…

2-Chloroethyl toluene-4-sulphonate 97%

Dodavatel: Apollo Scientific

Synonyms: 2-Chloroethyl p-toluenesulphonate

OR480570-100G
APOSOR480570-100GEA 4600 CZK
APOSOR480570-100G
2-Chloroethyl toluene-4-sulphonate 97%
2-Chloroethyl toluene-4-sulphonate
Vzorec: C₉H₁₁ClO₃S
MW: 234,7 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000970
CAS číslo: 80-41-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR