Tisk…

5-Bromo-2-chlorobenzothiazole 95%

Dodavatel: Apollo Scientific

Synonyms: 5-Bromo-2-chlorobenzo[d]thiazole

OR480532-1G OR480532-5G
APOSOR480532-1GEA 2240 CZK
APOSOR480532-1G APOSOR480532-5G
5-Bromo-2-chlorobenzothiazole 95%
5-Bromo-2-chlorobenzothiazole
Vzorec: C₇H₃BrClNS
MW: 248,53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09749227
CAS číslo: 824403-26-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR