2-Chlorobenzothiazole-6-carbonitrile 95%
Dodavatel: Apollo Scientific
OR480496-1G
OR480496-250MG
APOSOR480496-1GEA
4470
CZK
APOSOR480496-1G
APOSOR480496-250MG
2-Chlorobenzothiazole-6-carbonitrile 95%
2-Chlorobenzothiazole-6-carbonitrile
Vzorec:
C₈H₃ClN₂S MW: 194,6408 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09743933 CAS číslo: 80945-83-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...