2-Amino-4,4-dimethylpentanoic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
H-DL-3-tBu-Ala-OH, H-β-tBu-DL-Ala-OH
OR480463-1G
OR480463-5G
APOSOR480463-1GEA
4470
CZK
APOSOR480463-1G
APOSOR480463-5G
2-Amino-4,4-dimethylpentanoic acid 98%
2-Amino-4,4-dimethylpentanoic acid
Vzorec:
C₇H₁₅NO₂ MW: 145,202 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00066078 CAS číslo: 106247-35-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...