Tisk…

(H-Homocys-OH)₂

Dodavatel: Apollo Scientific
Varování

Synonyms: H-HoCystine-OH, (2S)-2-Amino-4-[[(3S)-3-amino-4-hydroxy-4-oxo-butyl]disulphanyl]butanoic acid, (H-HoCys-OH)2

OR480428-100G OR480428-25G
APOSOR480428-100GEA 8670 CZK
APOSOR480428-100G APOSOR480428-25G
(H-Homocys-OH)₂
(H-Homocys-OH)₂
Vzorec: C₈H₁₆N₂O₄S₂
Bod tání: 281…284 °C
Storage Temperature: Ambient
MDL Number: MFCD00020391
CAS číslo: 626-72-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR