Tisk…
1H-Pyrrolo[3,2-c]pyridine-6-carboxylic acid methyl ester 95%
Dodavatel: Apollo Scientific
Synonyms: Methyl 1H-pyrrolo[3,2-c]pyridine-6-carboxylate
OR480281-1G OR480281-250MG OR480281-500MG
APOSOR480281-1GEA 46620 CZK
APOSOR480281-1G APOSOR480281-250MG APOSOR480281-500MG
1H-Pyrrolo[3,2-c]pyridine-6-carboxylic acid methyl ester 95%
1H-Pyrrolo[3,2-c]pyridine-6-carboxylic acid methyl ester
Vzorec: C₉H₈N₂O₂
MW: 176,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18425635
CAS číslo: 1352394-18-3