Tisk…

(1-Methyl-1H-pyrazol-3-yl)acetic acid 95%

Dodavatel: Apollo Scientific

OR480274-1G OR480274-250MG OR480274-500MG
APOSOR480274-1GEA 16570 CZK
APOSOR480274-1G APOSOR480274-250MG APOSOR480274-500MG
(1-Methyl-1H-pyrazol-3-yl)acetic acid 95%
(1-Methyl-1H-pyrazol-3-yl)acetic acid
Vzorec: C₆H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD12028051
CAS číslo: 1177318-00-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR