6-Bromo[1,8]naphthyridin-2(1H)-one 98%
Dodavatel: Apollo Scientific
Synonyms:
6-Bromo-1,8-naphthyridin-2-ol
OR480268-1G
OR480268-500MG
OR480268-5G
APOSOR480268-1GEA
5660
CZK
APOSOR480268-1G
APOSOR480268-500MG
APOSOR480268-5G
6-Bromo[1,8]naphthyridin-2(1H)-one 98%
6-Bromo[1,8]naphthyridin-2(1H)-one
Vzorec:
C₈H₅BrN₂O MW: 225,05 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11226504 CAS číslo: 72754-05-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...