Tisk…

5-Bromo-3-iodo-6-methylpyridin-2-amine 95%

Dodavatel: Apollo Scientific

OR480261-10G OR480261-1G OR480261-5G
APOSOR480261-10GEA 18550 CZK
APOSOR480261-10G APOSOR480261-1G APOSOR480261-5G
5-Bromo-3-iodo-6-methylpyridin-2-amine 95%
5-Bromo-3-iodo-6-methylpyridin-2-amine
Vzorec: C₆H₆BrIN₂
Storage Temperature: Ambient
MDL Number: MFCD09880093
CAS číslo: 958357-86-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR