(3-Chloropyridin-4-yl)methanol 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR480260-1G
OR480260-250MG
OR480260-5G
APOSOR480260-1GEA
2820
CZK
APOSOR480260-1G
APOSOR480260-250MG
APOSOR480260-5G
(3-Chloropyridin-4-yl)methanol 98%
(3-Chloropyridin-4-yl)methanol
Vzorec:
C₆H₆ClNO Storage Temperature: Ambient |
MDL Number:
MFCD09910243 CAS číslo: 79698-53-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...