7-Methyl-1,2,3,4-tetrahydroisoquinoline 97%
Dodavatel: Apollo Scientific
Synonyms:
7-Methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride
OR480254-1G
OR480254-500MG
OR480254-5G
APOSOR480254-1GEA
8960
CZK
APOSOR480254-1G
APOSOR480254-500MG
APOSOR480254-5G
7-Methyl-1,2,3,4-tetrahydroisoquinoline 97%
7-Methyl-1,2,3,4-tetrahydroisoquinoline
Vzorec:
C₁₀H₁₃N MW: 147,2200 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03004622 CAS číslo: 207451-81-8 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...