Tisk…

4,6-Dihydro-1H-pyrrolo[3,4-c]pyrazole-5-carboxylic acid tert-butyl ester 95%

Dodavatel: Apollo Scientific

Synonyms: tert-Butyl 4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxylate

OR480248-1G OR480248-250MG OR480248-500MG
APOSOR480248-1GEA 13260 CZK
APOSOR480248-1G APOSOR480248-250MG APOSOR480248-500MG
4,6-Dihydro-1H-pyrrolo[3,4-c]pyrazole-5-carboxylic acid tert-butyl ester 95%
4,6-Dihydro-1H-pyrrolo[3,4-c]pyrazole-5-carboxylic acid tert-butyl ester
Vzorec: C₁₀H₁₅N₃O₂
MW: 209,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061257
CAS číslo: 657428-42-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR