(1H-Indol-4-ylmethyl)amine 97%
Dodavatel: Apollo Scientific
Synonyms:
4-Aminomethylindole, (1H-Indol-4-yl)methanamine, 4-(Aminomethyl)indole
OR480231-25G
OR480231-5G
APOSOR480231-25GEA
29920
CZK
APOSOR480231-25G
APOSOR480231-5G
(1H-Indol-4-ylmethyl)amine 97%
(1H-Indol-4-ylmethyl)amine
Vzorec:
C₉H₁₀N₂ MW: 146,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04973297 CAS číslo: 3468-18-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...