Tisk…

4-(Hydroxymethyl)-2-methylaniline 97%

Dodavatel: Apollo Scientific

OR480229-1G OR480229-5G
APOSOR480229-1GEA 4340 CZK
APOSOR480229-1G APOSOR480229-5G
4-(Hydroxymethyl)-2-methylaniline 97%
4-(Hydroxymethyl)-2-methylaniline
Vzorec: C₈H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD04038446
CAS číslo: 88990-57-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR