2-Chloro-6-hydroxybenzonitrile 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR480219-1G
OR480219-25G
OR480219-5G
APOSOR480219-1GEA
7970
CZK
APOSOR480219-1G
APOSOR480219-25G
APOSOR480219-5G
2-Chloro-6-hydroxybenzonitrile 97%
2-Chloro-6-hydroxybenzonitrile
Vzorec:
C₇H₄ClNO MW: 153,5658 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01646165 CAS číslo: 89999-90-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...