Tisk…

4-Amino-1-boc-piperidine-4-carboxylic acid 97%

Dodavatel: Apollo Scientific

OR480215-25G OR480215-5G
APOSOR480215-25GEA 8440 CZK
APOSOR480215-25G APOSOR480215-5G
4-Amino-1-boc-piperidine-4-carboxylic acid 97%
4-Amino-1-boc-piperidine-4-carboxylic acid
Vzorec: C₁₁H₂₀N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD01318728
CAS číslo: 183673-71-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR