4-Cyano-5-methylsulphanylthiophene-2-carboxylic acid methyl ester 97%
Dodavatel: Apollo Scientific
OR480203-1G
OR480203-5G
APOSOR480203-1GEA
5000
CZK
APOSOR480203-1G
APOSOR480203-5G
4-Cyano-5-methylsulphanylthiophene-2-carboxylic acid methyl ester 97%
4-Cyano-5-methylsulphanylthiophene-2-carboxylic acid methyl ester
Vzorec:
C₈H₇NO₂S₂ Storage Temperature: Ambient |
MDL Number:
MFCD00173757 CAS číslo: 175202-48-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...