1H-Indazole-4-carbonitrile
Dodavatel: Apollo Scientific
|
Varování
|
OR48010-1G
OR48010-500MG
APOSOR48010-1GEA
9760
CZK
APOSOR48010-1G
APOSOR48010-500MG
1H-Indazole-4-carbonitrile
1H-Indazole-4-carbonitrile
Vzorec:
C₈H₅N₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD09261250 CAS číslo: 861340-10-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...