Tisk…

4-(Bromomethyl)phenylboronic acid

Dodavatel: Apollo Scientific
Varování

OR4799-1G OR4799-5G OR4799-25G
APOSOR4799-1GEA 622 CZK
APOSOR4799-1G APOSOR4799-5G APOSOR4799-25G
4-(Bromomethyl)phenylboronic acid
4-(Bromomethyl)phenylboronic acid
Vzorec: C₇H₈BBrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD01318113
CAS číslo: 68162-47-0

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR