Ethyl benzoylacetate 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Ethyl 3-oxo-3-phenylpropanoate
OR4787-100G
OR4787-25G
APOSOR4787-100GEA
1620
CZK
APOSOR4787-100G
APOSOR4787-25G
Ethyl benzoylacetate 95%
Ethyl benzoylacetate
Vzorec:
C₁₁H₁₂O₃ MW: 192,21 g/mol Bod varu: 265…270 °C Hustota: 1,113 g/cm³ (25 °C) Bod vzplanutí: 140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009196 CAS číslo: 94-02-0 EINECS: 202-295-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...