Tisk…

4-Methoxy-3-pyridineboronic acid hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: (4-Methoxypyridin-3-yl)boronic acid hydrochloride

OR4783-250MG OR4783-5G
APOSOR4783-250MGEA 830 CZK
APOSOR4783-250MG APOSOR4783-5G
4-Methoxy-3-pyridineboronic acid hydrochloride
4-Methoxy-3-pyridineboronic acid hydrochloride
Vzorec: C₆H₉BClNO₃
MW: 189,4046 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04113800
CAS číslo: 874959-97-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR