Tisk…

1H-Pyrrolo[2,3-d]pyrimidin-4(7H)-one 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 7-Deazahypoxanthine

OR47823-1G OR47823-5G OR47823-25G
APOSOR47823-1GEA 622 CZK
APOSOR47823-1G APOSOR47823-5G APOSOR47823-25G
1H-Pyrrolo[2,3-d]pyrimidin-4(7H)-one 97%
1H-Pyrrolo[2,3-d]pyrimidin-4(7H)-one
Vzorec: C₆H₅N₃O
MW: 135.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08234923
CAS číslo: 3680-71-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR