5-Chloronicotinic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR4770-1G
OR4770-5G
APOSOR4770-1GEA
622
CZK
APOSOR4770-1G
APOSOR4770-5G
5-Chloronicotinic acid
5-Chloronicotinic acid
Vzorec:
C₆H₄ClNO₂ MW: 157.56 Bod tání: 169…171 °C Storage Temperature: Ambient |
MDL Number:
MFCD00128901 CAS číslo: 22620-27-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...