4-Hydroxypicolinic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Hydroxypicolinic acid, 4-Hydroxy-2-pyridinecarboxylic acid, 4-Hydroxypyridine-2-carboxylic acid
OR4750-1G
OR4750-250MG
APOSOR4750-1GEA
1120
CZK
APOSOR4750-1G
APOSOR4750-250MG
4-Hydroxypicolinic acid
4-Hydroxypicolinic acid
Vzorec:
C₆H₅NO₃ MW: 139.11 Storage Temperature: Ambient |
MDL Number:
MFCD00521155 CAS číslo: 22468-26-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...