Tisk…

2,5-Diamino-4,6-dichloropyrimidine

Dodavatel: Apollo Scientific
Varování

Synonyms: 4,6-Dichloropyrimidine-2,5-diamine

OR4725-1G OR4725-5G OR4725-25G
APOSOR4725-1GEA 622 CZK
APOSOR4725-1G APOSOR4725-5G APOSOR4725-25G
2,5-Diamino-4,6-dichloropyrimidine
2,5-Diamino-4,6-dichloropyrimidine
Vzorec: C₄H₄Cl₂N₄
MW: 179,0100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00792652
CAS číslo: 55583-59-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR