Tisk…

(3-Isopropyl-1,2,4-oxadiazol-5-yl)methanol 95%

Dodavatel: Apollo Scientific

OR471710-1G OR471710-250MG
APOSOR471710-1GEA 21460 CZK
APOSOR471710-1G APOSOR471710-250MG
(3-Isopropyl-1,2,4-oxadiazol-5-yl)methanol 95%
(3-Isopropyl-1,2,4-oxadiazol-5-yl)methanol
Vzorec: C₆H₁₀N₂O₂
Storage Temperature: Ambient
CAS číslo: 915925-45-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR