Tisk…

1-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 98%

Dodavatel: Apollo Scientific

Synonyms: 1-Methylpyrazole-3-boronic acid pinacol ester

OR471694-5G
APOSOR471694-5GEA 2940 CZK
APOSOR471694-5G
1-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 98%
1-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Vzorec: C₁₀H₁₇BN₂O₂
MW: 208.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114000
CAS číslo: 1020174-04-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR