Tisk…

(2-Piperidinophenyl)methanol 95%

Dodavatel: Apollo Scientific

Synonyms: 2-(1-Piperidinyl)benzyl alcohol

OR471676-1G OR471676-5G
APOSOR471676-1GEA 3940 CZK
APOSOR471676-1G APOSOR471676-5G
(2-Piperidinophenyl)methanol 95%
(2-Piperidinophenyl)methanol
Vzorec: C₁₂H₁₇NO
MW: 191,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05864821
CAS číslo: 87066-94-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR