Tisk…

6-Methyl-1H-indole-3-carboxylic acid 98%

Dodavatel: Apollo Scientific

OR471658-1G OR471658-5G
APOSOR471658-1GEA 1870 CZK
APOSOR471658-1G APOSOR471658-5G
6-Methyl-1H-indole-3-carboxylic acid 98%
6-Methyl-1H-indole-3-carboxylic acid
Vzorec: C₁₀H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD06203633
CAS číslo: 209920-43-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR