Tisk…

Benzyl-(S)-(-)-lactate 98%

Dodavatel: Apollo Scientific

Synonyms: Benzyl L-lactate, (S)-(-)-Lactic Acid Benzyl Ester, Benzyl (S)-Lactate

OR471654-25G
APOSOR471654-25GEA 4300 CZK
APOSOR471654-25G
Benzyl-(S)-(-)-lactate 98%
Benzyl-(S)-(-)-lactate
Vzorec: C₁₀H₁₂O₃
MW: 180,20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00209546
CAS číslo: 56777-24-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR