Tisk…

5-Methyl-1-Indanone 97%

Dodavatel: Apollo Scientific

Synonyms: 5-Methyl-1-indanone

OR471644-25G
APOSOR471644-25GEA 3910 CZK
APOSOR471644-25G
5-Methyl-1-Indanone 97%
5-Methyl-1-Indanone
Vzorec: C₁₀H₁₀O
MW: 146,189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179276
CAS číslo: 4593-38-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR