Tisk…

N-Acetyl-S-trityl-L-cysteine 95%

Dodavatel: Apollo Scientific

Synonyms: Ac-Cys(Trt)-OH

OR471635-100G OR471635-25G
APOSOR471635-100GEA 6390 CZK
APOSOR471635-100G APOSOR471635-25G
N-Acetyl-S-trityl-L-cysteine 95%
N-Acetyl-S-trityl-L-cysteine
Vzorec: C₂₄H₂₃NO₃S
MW: 405,512 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236747
CAS číslo: 27486-87-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR