2-Chloro-4,6-diphenyl-1,3,5-triazine 98%
Dodavatel: Apollo Scientific
OR471629-100G
APOSOR471629-100GEA
4570
CZK
APOSOR471629-100G
2-Chloro-4,6-diphenyl-1,3,5-triazine 98%
2-Chloro-4,6-diphenyl-1,3,5-triazine
Vzorec:
C₁₅H₁₀ClN₃ MW: 267,72 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05738885 CAS číslo: 3842-55-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...