Tisk…

Fmoc-N-methyl-D-valine 97%

Dodavatel: Apollo Scientific

Synonyms: N-Fmoc-N-methyl-D-valine, Fmoc-D-N-Me-Val-OH

OR471628-10G OR471628-25G OR471628-5G
APOSOR471628-10GEA 3190 CZK
APOSOR471628-10G APOSOR471628-25G APOSOR471628-5G
Fmoc-N-methyl-D-valine 97%
Fmoc-N-methyl-D-valine
Vzorec: C₂₁H₂₃NO₄
MW: 353.418 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00153396
CAS číslo: 103478-58-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR