Tisk…

Monoethyl succinate 97%

Dodavatel: Apollo Scientific

OR471619-100G OR471619-500G
APOSOR471619-100GEA 3310 CZK
APOSOR471619-100G APOSOR471619-500G
Monoethyl succinate 97%
Monoethyl succinate
Vzorec: C₆H₁₀O₄
Bod varu: 248 °C
Bod tání: 8 °C
Hustota: 1,141 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00053346
CAS číslo: 1070-34-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR