Tisk…

2-(2-Propynyloxy)tetrahydro-2H-pyran 99%

Dodavatel: Apollo Scientific
Varování

Synonyms: Tetrahydro-2-(2-propynyloxy)-2H-pyran, 2-Propargyloxane, 2-(Prop-2-yn-1-yloxy)tetrahydro-2H-pyran, Propargyl 2-tetrahydropyranyl ether, 2-(2-Propynyloxy)tetrahydropyran

OR471618-100G OR471618-25G OR471618-5G
APOSOR471618-100GEA 3740 CZK
APOSOR471618-100G APOSOR471618-25G APOSOR471618-5G
2-(2-Propynyloxy)tetrahydro-2H-pyran 99%
2-(2-Propynyloxy)tetrahydro-2H-pyran
Vzorec: C₈H₁₂O₂
MW: 140,18 g/mol
Bod varu: 65 °C
Hustota: 1,01 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00006604
CAS číslo: 6089-04-9
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR