Tisk…

[2-(1H-Pyrazol-3-yl)ethyl]amine dihydrochloride

Dodavatel: Apollo Scientific

OR471576-1G OR471576-5G
APOSOR471576-1GEA 13230 CZK
APOSOR471576-1G APOSOR471576-5G
[2-(1H-Pyrazol-3-yl)ethyl]amine dihydrochloride
[2-(1H-Pyrazol-3-yl)ethyl]amine dihydrochloride
Vzorec: C₅H₁₁Cl₂N₃
Storage Temperature: Ambient
MDL Number: MFCD01693704
CAS číslo: 138-92-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR