2,4,7-Trichloropyrido[2,3-d]pyrimidine
Dodavatel: Apollo Scientific
OR471510-1G
OR471510-250MG
APOSOR471510-1GEA
7280
CZK
APOSOR471510-1G
APOSOR471510-250MG
2,4,7-Trichloropyrido[2,3-d]pyrimidine
2,4,7-Trichloropyrido[2,3-d]pyrimidine
Vzorec:
C₇H₂Cl₃N₃ Storage Temperature: Ambient |
MDL Number:
MFCD11109463 CAS číslo: 938443-20-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...