Tisk…

Tetrakis(4-bromophenyl)methane

Dodavatel: Apollo Scientific

OR471493-5G
APOSOR471493-5GEA 6420 CZK
APOSOR471493-5G
Tetrakis(4-bromophenyl)methane
Tetrakis(4-bromophenyl)methane
Vzorec: C₂₅H₁₆Br₄
Storage Temperature: Ambient
MDL Number: MFCD19688472
CAS číslo: 105309-59-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR