Tisk…

1H-Indol-6-ylmethylamine 96%

Dodavatel: Apollo Scientific

Synonyms: (1H-Indol-6-yl)methanamine, 6-(Aminomethyl)indole

OR471270-5G OR471270-1G OR471270-25G
APOSOR471270-5GEA 9100 CZK
APOSOR471270-5G APOSOR471270-1G APOSOR471270-25G
1H-Indol-6-ylmethylamine 96%
1H-Indol-6-ylmethylamine
Vzorec: C₉H₁₀N₂
MW: 146,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06213839
CAS číslo: 3468-17-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR