Ethyl-2,4-dioxo-4-(2-pyridinyl)butanoate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Ethyl 2,4-dioxo-4-(pyridin-2-yl)butanoate
OR470834-5G
OR470834-1G
APOSOR470834-5GEA
15310
CZK
APOSOR470834-5G
APOSOR470834-1G
Ethyl-2,4-dioxo-4-(2-pyridinyl)butanoate
Ethyl-2,4-dioxo-4-(2-pyridinyl)butanoate
Vzorec:
C₁₁H₁₁NO₄ MW: 221,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08056636 CAS číslo: 92288-93-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...