Tisk…

[4-(tert-Butoxycarbonyl)morpholin-2-yl]acetic acid 96%

Dodavatel: Apollo Scientific

Synonyms: N-Boc-2-(2-morpholinyl)acetic acid

OR470743-5G OR470743-1G
APOSOR470743-5GEA 31610 CZK
APOSOR470743-5G APOSOR470743-1G
[4-(tert-Butoxycarbonyl)morpholin-2-yl]acetic acid 96%
[4-(tert-Butoxycarbonyl)morpholin-2-yl]acetic acid
Vzorec: C₁₁H₁₉NO₅
MW: 245,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114894
CAS číslo: 766539-28-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR