Tisk…

3-(3-Methoxyphenyl)propionaldehyde

Dodavatel: Apollo Scientific

OR470472-1G OR470472-250MG
APOSOR470472-1GEA 15970 CZK
APOSOR470472-1G APOSOR470472-250MG
3-(3-Methoxyphenyl)propionaldehyde
3-(3-Methoxyphenyl)propionaldehyde
Vzorec: C₁₀H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD07772909
CAS číslo: 40138-66-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR