Tisk…

4-Methyl[1,3,5]triazino[1,2-a]benzimidazol-2-amine

Dodavatel: Apollo Scientific

OR470440-1G OR470440-5G
APOSOR470440-1GEA 6980 CZK
APOSOR470440-1G APOSOR470440-5G
4-Methyl[1,3,5]triazino[1,2-a]benzimidazol-2-amine
4-Methyl[1,3,5]triazino[1,2-a]benzimidazol-2-amine
Vzorec: C₁₀H₉N₅
MW: 199.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04155098
CAS číslo: 78650-16-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR