Tisk…

6-Oxo-1,6-dihydropyridazine-3-carboxamide

Dodavatel: Apollo Scientific

Synonyms: 6-Hydroxypyridazine-3-carboxamide

OR470417-1G OR470417-5G
APOSOR470417-1GEA 8700 CZK
APOSOR470417-1G APOSOR470417-5G
6-Oxo-1,6-dihydropyridazine-3-carboxamide
6-Oxo-1,6-dihydropyridazine-3-carboxamide
Vzorec: C₅H₅N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD11643204
CAS číslo: 60184-73-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR