Methyl-3-nitropicolinate
Dodavatel: Apollo Scientific
Synonyms:
Methyl 3-Nitropyridine-2-carboxylate
OR470250-1G
OR470250-5G
APOSOR470250-1GEA
5530
CZK
APOSOR470250-1G
APOSOR470250-5G
Methyl-3-nitropicolinate
Methyl-3-nitropicolinate
Vzorec:
C₇H₆N₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD09953158 CAS číslo: 103698-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...