Tisk…

3-(3,4-Diethoxyphenyl)isoxazol-5-amine

Dodavatel: Apollo Scientific

Synonyms: 5-Amino-3-(3,4-diethoxyphenyl)isoxazole

OR470200-1G OR470200-5G
APOSOR470200-1GEA 8700 CZK
APOSOR470200-1G APOSOR470200-5G
3-(3,4-Diethoxyphenyl)isoxazol-5-amine
3-(3,4-Diethoxyphenyl)isoxazol-5-amine
Vzorec: C₁₃H₁₆N₂O₃
MW: 248,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07382988
CAS číslo: 501325-89-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR