Tisk…

Methyl-4-iodsalicylate 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Methyl 2-hydroxy-4-iodobenzoate

OR4702-100G OR4702-25G OR4702-5G
APOSOR4702-100GEA 6520 CZK
APOSOR4702-100G APOSOR4702-25G APOSOR4702-5G
Methyl-4-iodsalicylate 98%
Methyl-4-iodsalicylate
Vzorec: C₈H₇IO₃
MW: 278,05 g/mol
Bod tání: 68…73 °C
Storage Temperature: Ambient
MDL Number: MFCD06797864
CAS číslo: 18179-39-0
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR